Diphenylsilane, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
M-Clarity™ quality level = MQ100
148482-25G
148482-5G
SIAL148482-25GEA
3520
CZK
SIAL148482-25G
SIAL148482-5G
Diphenylsilane, Sigma-Aldrich®
Diphenylsilane
Vzorec:
C₁₂H₁₂Si Bod varu: 95 °C (1013 hPa) Hustota: 0,9962 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003002 CAS číslo: 775-12-2 EINECS: 212-271-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...