Tisk…

Trichloro(phenyl)silane, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: Phenyltrichlorosilane

M-Clarity™ quality level = MQ100

440108-100ML 440108-1L
SIAL440108-100MLEA 1130 CZK
SIAL440108-100ML SIAL440108-1L
Trichloro(phenyl)silane, Sigma-Aldrich®
Trichloro(phenyl)silane
Vzorec: C₆H₅Cl₃Si
Bod varu: 201 °C (1013 hPa)
Bod tání: –40 °C
Hustota: 1,315 g/cm³ (20 °C)
MDL Number: MFCD00000479
CAS číslo: 98-13-5
EINECS: 202-640-8
UN: 1804
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR