Trichloro(phenyl)silane ≥97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Phenyltrichlorosilane
A16713.22
A16713.36
A16713.22EA
1230
CZK
A16713.22
A16713.36
Trichloro(phenyl)silane ≥97%
Trichloro(phenyl)silane
Purity: 97%
CAS No.: 98-13-5
Molecular Formula: C6H5Cl3Si
Formula Weight: 211.55
Flash Point: 91°(195°F)
Refractive Index: 1.5230
Odour: Odourless
Melting Point: -127°
Boiling Point: 201°
Beilstein Reference: 508730
UN No.: 1804
MDL No.: MFCD00000479
Vzorec:
C₆H₅Cl₃Si MW: 211.55 Bod varu: 201 °C Bod tání: –127 °C Hustota: 1.324 Bod vzplanutí: 91 °C (195 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000479 CAS číslo: 98-13-5 EINECS: 202-640-8 UN: 1804 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...