Trichloro(phenyl)silane 95%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Phenyltrichlorosilane
131000100
131002500
131000010
ACRO131000100EA
780
CZK
ACRO131000100
ACRO131002500
ACRO131000010
Trichloro(phenyl)silane 95%
Trichloro(phenyl)silane
Vzorec:
C₆H₅Cl₃Si Bod varu: 201 °C (760 mmHg) Bod tání: –40 °C Hustota: 1,321 g/cm³ Bod vzplanutí: 91 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000479 CAS číslo: 98-13-5 EINECS: 202-640-8 UN: 1804 ADR: 8,II |
Výsledky specifikačního testu
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5220 to 1.5240 (20°C, 589 nm) |
GC | ≥94 % |
Hydroxyl value | 65 to 78 mg KOH/g |
Iodine value | 25 to 35 g I2/100 g |
Saponification value | 65 to 70 mg KOH/g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...