Hexamethyldisilane, Sigma-Aldrich®
Dodavatel: Merck
MQ100
217069-10G
217069-50G
217069-5G
SIAL217069-10GEA
2200
CZK
SIAL217069-10G
SIAL217069-50G
SIAL217069-5G
Hexamethyldisilane, Sigma-Aldrich®
Hexamethyldisilane
Vzorec:
C₆H₁₈Si₂ MW: 146.38 Bod varu: 112 Bod tání: 13…15 °C Hustota: 0.729 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00008258 CAS číslo: 1450-14-2 EINECS: 215-911-0 UN: 1993 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...