Tisk…

Hexabutylditin ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A12007.18 A12007.14 A12007.06 A12007.09 A12007.30
A12007.18EA 5980 CZK
A12007.18 A12007.14 A12007.06 A12007.09 A12007.30
Hexabutylditin ≥98%
Hexabutylditin

Purity: 98% CAS No.: 813-19-4 Molecular Formula: C24H54Sn2 Formula Weight: 580.07 Storage Temperature: -30°C to -10°C Flash Point: 124°(255°F) Refractive Index: 1.5120 Boiling Point: 197-198°/10mm Beilstein Reference: 3605476 UN No.: 2788 MDL No.: MFCD00009417

Vzorec: C₂₄H₅₄Sn₂
Bod varu: 197…198 °C (10 mmHg)
Hustota: 1,152
Bod vzplanutí: 124 °C (255 °F)
Storage Temperature: Freezer
MDL Number: MFCD00009417
CAS číslo: 813-19-4
EINECS: 212-383-3
UN: 2788
ADR: 6.1,III

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR