Triethylsilane, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
230197-25G
230197-5G
230197-100G
SIAL230197-25GEA
1930
CZK
SIAL230197-25G
SIAL230197-5G
SIAL230197-100G
Triethylsilane, Sigma-Aldrich®
Triethylsilane
Vzorec:
C₆H₁₆Si Bod varu: 107 °C (1013 hPa) Hustota: 0,73 g/cm³ (20 °C) Bod vzplanutí: –3 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00009018 CAS číslo: 617-86-7 EINECS: 210-535-3 UN: 1993 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...