Chlortrimethylsilan, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
TMCS, TMSC, Trimethylchlorosilane
M-Clarity™ quality level = MQ200
386529-100ML
386529-1L
SIAL386529-100MLEA
2590
CZK
SIAL386529-100ML
SIAL386529-1L
Chlortrimethylsilan, Sigma-Aldrich®
Chlortrimethylsilan
Vzorec:
C₃H₉ClSi MW: 108,64 g/mol Bod varu: 59 °C (1013 hPa) Bod tání: –58 °C Hustota: 0,8576 g/cm³ (20 °C) Bod vzplanutí: –18 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000502 CAS číslo: 75-77-4 EINECS: 200-900-5 UN: 1298 ADR: 3,II REACH: 01-2119457596-25 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...