Tributyltin chloride, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
Chlorotributylstannane, Tri-n-butyltin chloride, TBTC
M-Clarity™ quality level = MQ100
T50202-100G
T50202-500G
T50202-5G
SIALT50202-100GEA
1730
CZK
SIALT50202-100G
SIALT50202-500G
SIALT50202-5G
Tributyltin chloride, Sigma-Aldrich®
Tributyltin chloride
Vzorec:
C₁₂H₂₇ClSn Bod varu: 171…173 °C (25 mmHg) Bod tání: –9 °C Hustota: 1,2 g/cm³ (20 °C) Bod vzplanutí: >100 °C |
MDL Number:
MFCD00000521 CAS číslo: 1461-22-9 EINECS: 215-958-7 UN: 2788 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...