Tisk…

Dioctyltin oxide

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Di-n-octyltin oxide, Dioctyloxostannane

71193.14 71193.22 71193.36
71193.14EA 937 CZK
71193.14 71193.22 71193.36
Dioctyltin oxide
Dioctyltin oxide

CAS No.: 870-08-6 Molecular Formula: (C8H17)2SnO Formula Weight: 361.14 Physical Form: Powder Appearance: White Melting Point: 245-248° dec. MDL No.: MFCD00013839

Vzorec: C₁₆H₃₄OSn
MW: 361.16
Bod varu: 230
Bod tání: 245…248 °C (rozkl.)
Hustota: 1.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013839
CAS číslo: 870-08-6
EINECS: 212-791-1
UN: 3146
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR