Tisk…

Dimethyltin dichloride ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A17661.06 A17661.14
A17661.06EA 2320 CZK
A17661.06 A17661.14
Dimethyltin dichloride ≥98%
Dimethyltin dichloride

Purity: 98% CAS No.: 753-73-1 Molecular Formula: (CH3)2SnCl2 Formula Weight: 219.69 Melting Point: 104-108° Boiling Point: 188-190° UN No.: 3146 MDL No.: MFCD00000517

Vzorec: C₂H₆Cl₂Sn
Bod varu: 188…190 °C
Bod tání: 104…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00000517
CAS číslo: 753-73-1
EINECS: 212-039-2
UN: 3146
ADR: 6.1,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR