Tisk…

Dimethyldichlorsilan, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: Dichloro(dimethyl)silane, Dichlorodimethylsilane

M-Clarity™ quality level = MQ200

440272-100ML 440272-1L
SIAL440272-100MLEA 1430 CZK
SIAL440272-100ML SIAL440272-1L
Dimethyldichlorsilan, Sigma-Aldrich®
Dimethyldichlorsilan
Vzorec: C₂H₆Cl₂Si
MW: 129.06
Bod varu: 70
Bod tání: –76 °C
Hustota: 1.0745 g/cm³ (20 °C)
Bod vzplanutí: –16 °C
MDL Number: MFCD00000491
CAS číslo: 75-78-5
EINECS: 200-901-0
UN: 1162
ADR: 3 II
REACH: 01-2119437250-51

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR