Chlorodimethylsilane 97%, rods
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Dimethylchlorosilane
A13113.AB
A13113.AD
A13113.AK
A13113.ABEA
913
CZK
A13113.AB
A13113.AD
A13113.AK
Chlorodimethylsilane 97%, rods
Chlorodimethylsilane
Purity: 97%
CAS No.: 1066-35-9
Molecular Formula: C2H7ClSi
Formula Weight: 94.62
Storage Temperature: 2°C to 8°C
Flash Point: -28°(-18°F)
Refractive Index: 1.3830
Odour: Acrid
Melting Point: -111°
Boiling Point: 34-36°
Beilstein Reference: 1633434
UN No.: 2924
MDL No.: MFCD00000495
Vzorec:
C₂H₇ClSi MW: 94.62 Bod varu: 34…36 °C Bod tání: –111 °C Hustota: 0.869 Bod vzplanutí: –28 °C (–18 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000495 CAS číslo: 1066-35-9 EINECS: 213-912-0 UN: 2924 ADR: 3 I REACH: 01-2119486782-25 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...