(3-Mercaptopropyl)triethoxysilane, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
3-(Triethoxysilyl)propanethiol, 3-Mercaptopropyltriethoxysilane
63797-100ML-F
63797-25ML-F
SIAL63797-100ML-FEA
11400
CZK
SIAL63797-100ML-F
SIAL63797-25ML-F
(3-Mercaptopropyl)triethoxysilane, Sigma-Aldrich®
(3-Mercaptopropyl)triethoxysilane
Vzorec:
HSH₂CCH₂CH₂Si(OCH₂CH₃)₃ MW: 238,42 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00053924 CAS číslo: 14814-09-6 EINECS: 238-883-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...