Tisk…
Methyltrimethoxysilane, Sigma-Aldrich®
Dodavatel: Merck
Synonyms: Trimethoxy(methyl)silane
M-Clarity™ quality level = MQ100
69471-1L 69471-250ML
SIAL69471-1LEA 5790 CZK
SIAL69471-1L SIAL69471-250ML
Methyltrimethoxysilane, Sigma-Aldrich®
Methyltrimethoxysilane
Vzorec: C₄H₁₂O₃Si
MW: 136.22
Bod varu: 102
Bod tání: 11
Hustota: 0.955 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008342
CAS číslo: 1185-55-3
EINECS: 214-685-0