Hexyltrimethoxysilane ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Trimethoxy(hexyl)silane
L03576.06
L03576.14
L03576.06EA
855
CZK
L03576.06
L03576.14
Hexyltrimethoxysilane ≥97%
Hexyltrimethoxysilane
Purity: 97%
CAS No.: 3069-19-0
Molecular Formula: CH3(CH2)5Si(OCH3)3
Formula Weight: 206.36
Flash Point: 62°(143°F)
Refractive Index: 1.4070
Boiling Point: 202-203°
Beilstein Reference: 2347551
MDL No.: MFCD00039801
Vzorec:
C₉H₂₂O₃Si MW: 206,36 g/mol Bod varu: 202…203 °C Hustota: 0,920 Bod vzplanutí: 62 °C (143 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00039801 CAS číslo: 3069-19-0 EINECS: 221-331-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...