Tisk…

Decyltrimethoxysilane ≥98%

Dodavatel: Thermo Fisher Scientific

L16966.14 L16966.22 AB171953.0025
L16966.14EA 1670 CZK
L16966.14 L16966.22 ABCRAB171953.0025
Decyltrimethoxysilane ≥98%
Decyltrimethoxysilane

Purity: 98% CAS No.: 5575-48-4 Molecular Formula: CH3(CH2)9Si(OCH3)3 Formula Weight: 262.47 Flash Point: 117°(242°F) Refractive Index: 1.4220 Melting Point: -37° Boiling Point: 102-106°/3.5mm Beilstein Reference: 8625082 MDL No.: MFCD00069135

Vzorec: C₁₃H₃₀O₃Si
Storage Temperature: Ambient
MDL Number: MFCD00069135
CAS číslo: 5575-48-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR