Tisk…

Isobutyltrimethoxysilane, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

444065-100ML 444065-25ML
SIAL444065-100MLEA 1890 CZK
SIAL444065-100ML SIAL444065-25ML
Isobutyltrimethoxysilane, Sigma-Aldrich®
Isobutyltrimethoxysilane
Vzorec: (H₃C)₂HCSi(OCH₃)₃
MW: 178.3
Bod varu: 155
Hustota: 0.933 g/cm³ (20 °C)
MDL Number: MFCD00051589
CAS číslo: 18395-30-7
EINECS: 242-272-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR