N-(3-(trimethoxysilyl)propyl)ethylendiamin 96%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
3-(2-Aminoethylamino)propyltrimethoxysilane, N-(2-Aminoethyl)-3-aminopropyltrimethoxysilane, N-[3-(Trimethoxysilyl)propyl]ethylenediamine
Purity: 96%
CAS No.: 1760-24-3
Molecular Formula: (CH3O)3Si(CH2)3NHCH2CH2NH2
Formula Weight: 222.36
Flash Point: 136°(276°F)
Refractive Index: 1.4435
Odour: Ammonia-like
Boiling Point: 261-263°
Beilstein Reference: 636230
UN No.: 2735
MDL No.: MFCD00008173
Vzorec:
H₂NCH₂CH₂NHCH₂CH₂CH₂Si(OCH₃)₃ MW: 222,36 g/mol Bod varu: 261…263 °C Bod tání: ∼ 0 °C Hustota: 1,010 Bod vzplanutí: 136 °C (276 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008173 CAS číslo: 1760-24-3 EINECS: 217-164-6 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...