Tisk…

(3-Iodopropyl)trimethoxysilane, Sigma-Aldrich®

Dodavatel: Merck

58035-100ML-F 58035-25ML-F
SIAL58035-100ML-FEA 12450 CZK
SIAL58035-100ML-F SIAL58035-25ML-F
(3-Iodopropyl)trimethoxysilane, Sigma-Aldrich®
(3-Iodopropyl)trimethoxysilane
Vzorec: IH₂CCH₂CH₂Si(OCH₃)₃
MDL Number: MFCD00054838
CAS číslo: 14867-28-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR