(3-Chloropropyl)trimethoxysilane, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
440183-100ML
440183-1L
SIAL440183-100MLEA
2640
CZK
SIAL440183-100ML
SIAL440183-1L
(3-Chloropropyl)trimethoxysilane, Sigma-Aldrich®
(3-Chloropropyl)trimethoxysilane
Vzorec:
ClH₂CCH₂CH₂Si(OCH₃)₃ Bod varu: 195 °C (1013 hPa) Bod tání: –50 °C Hustota: 1,082 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000997 CAS číslo: 2530-87-2 EINECS: 219-787-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...