Tisk…

Trichloro(methyl)germane ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Trichloro(methyl)germane, Methylgermanium trichloride

41449.02 41449.04 41449.09
41449.02EA 2180 CZK
41449.02 41449.04 41449.09
Trichloro(methyl)germane ≥97%
Trichloro(methyl)germane

Purity: 97% CAS No.: 993-10-2 Molecular Formula: CH3Cl3Ge Formula Weight: 193.99 Flash Point: 10°(50°F) Physical Form: Liquid Density: 1.706 Refractive Index: 1.4685 Sensitivity: Moisture Sensitive Appearance: Colourless to pale yellow Odour: Chlorine-like Boiling Point: 111° UN No.: 2924 MDL No.: MFCD00013585

Vzorec: CH₃Cl₃Ge
MW: 194
Bod varu: 111 °C
Hustota: 1.706
Bod vzplanutí: 10 °C (50 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013585
CAS číslo: 993-10-2
EINECS: 213-604-6
UN: 2924
ADR: 3 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR