Tisk…

Tetravinylsilane, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

318256-25G 318256-5G
SIAL318256-25GEA 13180 CZK
SIAL318256-25G SIAL318256-5G
Tetravinylsilane, Sigma-Aldrich®
Tetravinylsilane
Vzorec: C₈H₁₂Si
Bod varu: 125 °C (1013 hPa)
Hustota: 0,7984 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008607
CAS číslo: 1112-55-6
EINECS: 214-192-0
UN: 1993
ADR: 3,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR