Tisk…

Vinyltrimethoxysilane, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí


M-Clarity™ quality level = MQ200

440221-100ML 440221-1L
SIAL440221-100MLEA 3470 CZK
SIAL440221-100ML SIAL440221-1L
Vinyltrimethoxysilane, Sigma-Aldrich®
Vinyltrimethoxysilane
Vzorec: C₅H₁₂O₃Si
MW: 148,23 g/mol
Bod varu: 123 °C (1013 hPa)
Bod tání: 0 °C
Hustota: 0,97 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008605
CAS číslo: 2768-02-7
EINECS: 220-449-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR