Vinyltriethoxysilane
Dodavatel: abcr
|
Nebezpečí
|
Synonyms:
Triethoxy(vinyl)silane
AB111594.0025
AB111594.0100
ABCRAB111594.0025EA
0
CZK
ABCRAB111594.0025
ABCRAB111594.0100
Vinyltriethoxysilane
Vinyltriethoxysilane
Vzorec:
C₈H₁₈O₃Si MW: 190.31 Bod varu: 160 Hustota: 0.91 g/cm³ (20 °C) Bod vzplanutí: 34 Storage Temperature: Ambient |
MDL Number:
MFCD00009063 CAS číslo: 78-08-0 EINECS: 201-081-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...