Tisk…

Tetrapropoxysilane ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

L16644.14 L16644.22
L16644.14EA 1400 CZK
L16644.14 L16644.22
Tetrapropoxysilane ≥97%
Tetrapropoxysilane

Purity: 97% CAS No.: 682-01-9 Molecular Formula: C12H28O4Si Formula Weight: 264.44 Flash Point: 95°(203°F) Refractive Index: 1.4010 Boiling Point: 224-225° (94°/5mm) Beilstein Reference: 1706115 MDL No.: MFCD00009364

Vzorec: C₁₂H₂₈O₄Si
Bod varu: 224…225 °C/ 94 °C (5 mmHg)
Hustota: 0,916
Bod vzplanutí: 95 °C (203 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009364
CAS číslo: 682-01-9
EINECS: 211-659-0

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR