Tetramethoxysilane, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Tetramethyl orthosilicate
M-Clarity™ quality level = MQ200
87682-1L
87682-250ML
SIAL87682-1LEA
18530
CZK
SIAL87682-1L
SIAL87682-250ML
Tetramethoxysilane, Sigma-Aldrich®
Tetramethoxysilane
Vzorec:
C₄H₁₂O₄Si MW: 152.22 Bod varu: 121 Bod tání: –2 °C Hustota: 1.02 g/cm³ (20 °C) Bod vzplanutí: 26 Storage Temperature: Ambient |
MDL Number:
MFCD00008341 CAS číslo: 681-84-5 EINECS: 211-656-4 UN: 2606 ADR: 6.1 I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...