Tisk…

Tetramethoxysilane, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Tetramethyl orthosilicate

M-Clarity™ quality level = MQ200

341436-100G 341436-25G
SIAL341436-100GEA 4310 CZK
SIAL341436-100G SIAL341436-25G
Tetramethoxysilane, Sigma-Aldrich®
Tetramethoxysilane
Vzorec: C₄H₁₂O₄Si
MW: 152.22
Bod varu: 121
Bod tání: –2 °C
Hustota: 1.02 g/cm³ (20 °C)
Bod vzplanutí: 26
Storage Temperature: Ambient
MDL Number: MFCD00008341
CAS číslo: 681-84-5
EINECS: 211-656-4
UN: 2606
ADR: 6.1 I

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR