Tetraethoxysilan, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
Tetraethoxysilane
86578-1L
86578-250ML
SIAL86578-1LEA
3140
CZK
SIAL86578-1L
SIAL86578-250ML
Tetraethoxysilan, Sigma-Aldrich®
Tetraethyl orthosilicate (TEOS)
Vzorec:
Si(OC₂H₅)₄ Bod varu: 168 °C (1013 hPa) Hustota: 0,933 g/cm³ (20 °C) Bod vzplanutí: 38 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009062 CAS číslo: 78-10-4 EINECS: 201-083-8 UN: 1292 ADR: 3,III Merck Index: 13,03882 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...