Tetraethoxysilan ≥99.9%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Tetraethoxysilane
40251.22
40251.A3
40251.22EA
1820
CZK
40251.22
40251.A3
Tetraethoxysilan ≥99.9%
Tetraethyl orthosilicate (TEOS)
Purity: 99.9%
CAS No.: 78-10-4
Molecular Formula: C8H20O4Si
Formula Weight: 208.33
Flash Point: 46°(116°F)
Physical Form: Liquid
Density: 0.934
Refractive Index: 1.3820
Sensitivity: Moisture Sensitive
Appearance: Colourless
Odour: Pleasant
Melting Point: -85°
Boiling Point: 168°
Beilstein Reference: 1422225
Merck Reference: 14,3851
UN No.: 1292
MDL No.: MFCD00009062
Vzorec:
Si(OC₂H₅)₄ MW: 208,33 g/mol Bod varu: 168 °C Bod tání: –85 °C Hustota: 0,934 Bod vzplanutí: 46 °C (116 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009062 CAS číslo: 78-10-4 EINECS: 201-083-8 UN: 1292 ADR: 3,III Merck Index: 14,3851 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...