Tisk…

Germanium(IV) ethoxide, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Germanium tetraethanolate, Germanic acid tetraethyl ester, Tetraethyl orthogermanate, Germanium tetraethoxide, Germanium (IV) ethanolate

M-Clarity™ quality level = MQ100

339180-25G 339180-5G
SIAL339180-25GEA 29330 CZK
SIAL339180-25G SIAL339180-5G
Germanium(IV) ethoxide, Sigma-Aldrich®
Germanium(IV) ethoxide
Vzorec: C₈H₂₀GeO₄
MW: 252,85 g/mol
Bod varu: 185…187 °C (1013 hPa)
Bod tání: –72 °C
Hustota: 1,14 g/cm³ (20 °C)
MDL Number: MFCD00015121
CAS číslo: 14165-55-0
EINECS: 238-007-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR