Tisk…

(3-Acryloxypropyl)trimethoxysilane

Dodavatel: abcr

AB108961.0025 AB108961.0100 AB108961.1000 AB108961.2000
ABCRAB108961.0025EA 0 CZK
ABCRAB108961.0025 ABCRAB108961.0100 ABCRAB108961.1000 ABCRAB108961.2000
(3-Acryloxypropyl)trimethoxysilane
(3-Acryloxypropyl)trimethoxysilane
Vzorec: C₉H₁₈O₅Si
MDL Number: MFCD00054803
CAS číslo: 4369-14-6
EINECS: 419-560-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR