Triethoxysilane, Sigma-Aldrich®
Dodavatel: Merck
MQ200
390143-10ML
390143-50ML
SIAL390143-10MLEA
2360
CZK
SIAL390143-10ML
SIAL390143-50ML
Triethoxysilane, Sigma-Aldrich®
Triethoxysilane
Vzorec:
C₆H₁₆O₃Si MW: 164.28 Bod varu: 135 Bod tání: –170 °C Hustota: 0.8753 g/cm³ (20 °C) |
MDL Number:
MFCD00009061 CAS číslo: 998-30-1 EINECS: 213-650-7 UN: 3384 ADR: 6.1 I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...