Tisk…

Oxid ciničitý ≥99.996% (metals basis), Puratronic®

Dodavatel: Thermo Fisher Scientific

10891.06 10891.14
10891.06EA 3990 CZK
10891.06 10891.14
Oxid ciničitý ≥99.996% (metals basis), Puratronic®
Oxid ciničitý

Purity: 99.996% CAS No.: 18282-10-5 Molecular Formula: O2Sn Formula Weight: 150.69 Physical Form: -22 Mesh Powder Density: 6.95 Refractive Index: 1.9968 Appearance: White Odour: Odourless Melting Point: 1630° Boiling Point: 1800-1900° subl. Merck Reference: 14,8778 MDL No.: MFCD00011244

Vzorec: O₂Sn
Bod varu: 1800…1900 °C (subl.)
Bod tání: 1630 °C
Hustota: 6,95
Storage Temperature: Ambient
MDL Number: MFCD00011244
CAS číslo: 18282-10-5
EINECS: 242-159-0
Merck Index: 14,8778

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR