Silikagel, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide
M-Clarity™ quality level = MQ100
342890-100G
342890-1KG
SIAL342890-100GEA
3650
CZK
SIAL342890-100G
SIAL342890-1KG
Silikagel, Sigma-Aldrich®
Silicon dioxide
Vzorec:
SiO₂ MW: 60.08 Bod varu: 2230 Bod tání: 1719 Hustota: 2.2 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011232 CAS číslo: 7631-86-9 EINECS: 231-545-4 Merck Index: 14,08493 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...