Tisk…

Silicon(II) oxide, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Silicon monoxide, Silicon oxide

M-Clarity™ quality level = MQ100

262951-250G 262951-50G
SIAL262951-250GEA 19640 CZK
SIAL262951-250G SIAL262951-50G
Silicon(II) oxide, Sigma-Aldrich®
Silicon(II) oxide
Vzorec: SiO
Bod varu: 1880 °C (1013 hPa)
Bod tání: 1702 °C
Hustota: 2,13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00151536
CAS číslo: 10097-28-6
EINECS: 233-232-8
Merck Index: 12,08640

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR