Oxid germaničitý ≥99.99% (metals basis)
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Germanium dioxide
36382.04
36382.09
36382.18
36382.04EA
3090
CZK
36382.04
36382.09
36382.18
Oxid germaničitý ≥99.99% (metals basis)
Oxid germaničitý
Purity: 99.99%
CAS No.: 1310-53-8
Molecular Formula: GeO2
Formula Weight: 104.59
Physical Form: 3-12mm Sintered Lump
Density: 6.239
Appearance: White
Odour: Odourless
Melting Point: 1086°
Merck Reference: 14,4409
MDL No.: MFCD00011030
Used in phosphors, transistors and diodes, and infrared transmitting glass
Vzorec:
GeO₂ Bod tání: 1086 °C Hustota: 6,239 Storage Temperature: Ambient |
MDL Number:
MFCD00011030 CAS číslo: 1310-53-8 EINECS: 215-180-8 Merck Index: 14,4409 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...