Tisk…

Tin(IV) iodide, ultra dry ≥99.998% (metals basis)

Dodavatel: Thermo Fisher Scientific
Nebezpečí


Ampuled under argon

13617.06 13617.14
13617.06EA 7760 CZK
13617.06 13617.14
Tin(IV) iodide, ultra dry ≥99.998% (metals basis)
Tin(IV) iodide

Purity: 99.998% CAS No.: 7790-47-8 Molecular Formula: SnI4 Formula Weight: 626.31 Physical Form: Powder, Ampouled under argon Density: 4.46 Refractive Index: 2.106 Sensitivity: Moisture Sensitive Melting Point: ca 143° Boiling Point: 340° Merck Reference: 14,8777 UN No.: 3260 MDL No.: MFCD00036272

Vzorec: I₄Sn
Bod varu: 340 °C
Bod tání: ∼ 143 °C
Hustota: 4,46
Storage Temperature: Ambient
MDL Number: MFCD00036272
CAS číslo: 7790-47-8
EINECS: 232-208-4
UN: 3260
ADR: 8,III
Merck Index: 14,8777

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR