Tin(II) iodide ≥99%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
71112.09
71112.18
71112.09EA
4740
CZK
71112.09
71112.18
Tin(II) iodide ≥99%
Tin(II) iodide
Purity: 99+%
CAS No.: 10294-70-9
Molecular Formula: SnI2
Formula Weight: 372.50
Physical Form: -6 Mesh Powder
Density: 5.28
Appearance: Yellow to red
Melting Point: 320°
Boiling Point: 720° dec.
Solubility: Soluble in benzene, CS2, chloroform, and solutions of alkali chlorides or iodides. Decomposes in water
Merck Reference: 14,8785
UN No.: 3260
MDL No.: MFCD00049545
Vzorec:
I₂Sn MW: 372,52 g/mol Bod varu: 720 °C (rozkl.) Bod tání: 320 °C Hustota: 5,28 Storage Temperature: Ambient |
MDL Number:
MFCD00049545 CAS číslo: 10294-70-9 EINECS: 233-667-3 UN: 3260 ADR: 8,III Merck Index: 14,8785 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...