Germanium(IV) iodide ≥99.999% (metals basis)
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
14009.09
14009.18
14009.09EA
11430
CZK
14009.09
14009.18
Germanium(IV) iodide ≥99.999% (metals basis)
Germanium(IV) iodide
Purity: 99.999%
CAS No.: 13450-95-8
Molecular Formula: GeI4
Formula Weight: 580.21
Physical Form: -10 Mesh Crystalline
Density: 4.32
Melting Point: 146°
UN No.: 3260
MDL No.: MFCD00016120
Vzorec:
GeI₄ Bod varu: 350 °C (1013 hPa) Bod tání: 146 °C Hustota: 4,32 Storage Temperature: Ambient |
MDL Number:
MFCD00016120 CAS číslo: 13450-95-8 EINECS: 236-613-7 UN: 3260 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...