Lead(II) fluoride ≥99%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Lead difluoride
88282.30
88282.A1
88282.30EA
3290
CZK
88282.30
88282.A1
Lead(II) fluoride ≥99%
Lead(II) fluoride
Purity: 99% min
CAS No.: 7783-46-2
Molecular Formula: PbF2
Formula Weight: 245.20
Physical Form: Powder
Density: 8.445
Melting Point: 855°
Boiling Point: 1290°
Merck Reference: 14,5408
UN No.: 2291
MDL No.: MFCD00011162
Vzorec:
F₂Pb Bod varu: 1290 °C Bod tání: 855 °C Hustota: 8,445 Storage Temperature: Ambient |
MDL Number:
MFCD00011162 CAS číslo: 7783-46-2 EINECS: 231-998-8 UN: 2291 ADR: 6.1,III Merck Index: 14,5408 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...