Oxid olovnatý ≥99.9%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Lead monoxide
12220.36
12220.A3
12220.36EA
1720
CZK
12220.36
12220.A3
Oxid olovnatý ≥99.9%
Oxid olovnatý
Purity: 99.9%
CAS No.: 1317-36-8
Molecular Formula: PbO
Formula Weight: 223.19
Physical Form: -325 Mesh Powder
Density: 9.53
Melting Point: 886°
Merck Reference: 14,5413
UN No.: 3077
MDL No.: MFCD00011164
Vzorec:
PbO MW: 223,2 g/mol Bod varu: 1470 °C (1013 hPa) Bod tání: 886 °C Hustota: 9,53 Storage Temperature: Ambient |
MDL Number:
MFCD00011164 CAS číslo: 1317-36-8 EINECS: 215-267-0 UN: 3077 ADR: 9,III Merck Index: 14,5413 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...