Tisk…

Silicon carbide, β-phase, nanopowder

Dodavatel: Thermo Scientific
Varování

44646.06 44646.14 44646.22
44646.06EA 2480 CZK
44646.06 44646.14 44646.22
Silicon carbide, β-phase, nanopowder
Silicon carbide, β-phase

CAS No.: 409-21-2 Molecular Formula: SiC Formula Weight: 40.10 Physical Form: 45-55nm APS Powder, S.A. 70-90m2/g Odour: Odourless Melting Point: 2700° MDL No.: MFCD00049531

Vzorec: SiC
MW: 40,10 g/mol
Bod tání: 2700 °C
Storage Temperature: Ambient
MDL Number: MFCD00049531
CAS číslo: 409-21-2
EINECS: 206-991-8
Merck Index: 14,8492

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR