Silicon carbide, β-phase ≥95%, nanopowder
Dodavatel: Thermo Scientific
|
Varování
|
43884.14
43884.22
43884.06
43884.14EA
8480
CZK
43884.14
43884.22
43884.06
Silicon carbide, β-phase ≥95%, nanopowder
Silicon carbide, β-phase
Purity: 95% min
CAS No.: 409-21-2
Molecular Formula: SiC
Formula Weight: 40.10
Physical Form:
Vzorec:
SiC MW: 40,10 g/mol Bod tání: 2700 °C Storage Temperature: Ambient |
MDL Number:
MFCD00049531 CAS číslo: 409-21-2 EINECS: 206-991-8 Merck Index: 14,8492 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...