Karbid křemičitý ≥99% (metals basis)
Dodavatel: Thermo Fisher Scientific
|
Varování
|
43332.A3
43332.36
43332.A3EA
18430
CZK
43332.A3
43332.36
Karbid křemičitý ≥99% (metals basis)
Karbid křemičitý
Purity: 99%
CAS No.: 409-21-2
Molecular Formula: CSi
Formula Weight: 40.10
Physical Form: -325 Mesh Powder, Black
Density: 3.16
Appearance: Black
Odour: Odourless
Melting Point: 2700°
Merck Reference: 14,8492
MDL No.: MFCD00049531
Vzorec:
SiC MW: 40.1 Bod tání: 2700 Hustota: 3.16 Storage Temperature: Ambient |
MDL Number:
MFCD00049531 CAS číslo: 409-21-2 EINECS: 206-991-8 Merck Index: 14,8492 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...