Tisk…

Tin(IV) bromide ≥99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

71120.18 71120.30
71120.18EA 3680 CZK
71120.18 71120.30
Tin(IV) bromide ≥99%
Tin(IV) bromide

Purity: 99% CAS No.: 7789-67-5 Molecular Formula: SnBr4 Formula Weight: 438.33 Physical Form: Lump Density: 3.34 Appearance: Colourless to white Odour: Acidic Melting Point: 31° Boiling Point: 202°/734mm Merck Reference: 14,8773 UN No.: 3260 MDL No.: MFCD00011240

Vzorec: Br₄Sn
Bod varu: 202 °C (734 mmHg)
Bod tání: 31 °C
Hustota: 3,34
Storage Temperature: Ambient
MDL Number: MFCD00011240
CAS číslo: 7789-67-5
EINECS: 232-184-5
UN: 3260
ADR: 8,II
Merck Index: 14,8773

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR