Tisk…

Aluminium sulphate, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: Aluminum sulphate

M-Clarity™ quality level = MQ100

202614-100G 202614-25G 202614-5G
SIAL202614-100GEA 28840 CZK
SIAL202614-100G SIAL202614-25G SIAL202614-5G
Aluminium sulphate, Sigma-Aldrich®
Aluminium sulphate
Vzorec: Al₂O₁₂S₃
Bod tání: 770 °C
Hustota: 2,672 g/cm³ (22.5 °C)
MDL Number: MFCD00003423
CAS číslo: 10043-01-3
EINECS: 233-135-0
Merck Index: 12,00381

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR