Aluminium sulphate, anhydrous ≥99.99% (metals basis)
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Aluminum sulphate
Packaged under argon.
44563.06
44563.14
44563.22
44563.36
44563.06EA
1750
CZK
44563.06
44563.14
44563.22
44563.36
Aluminium sulphate, anhydrous ≥99.99% (metals basis)
Aluminium sulphate
Purity: 99.99%
CAS No.: 10043-01-3
Molecular Formula: Al2(SO4)3
Formula Weight: 342.15
Physical Form: Powder, Packaged under argon
Density: 2.71
pH: 3.5
Melting Point: 770°
Merck Reference: 14,366
MDL No.: MFCD00003423
Vzorec:
Al₂O₁₂S₃ MW: 342,15 g/mol Bod tání: 770 °C Hustota: 2,71 Storage Temperature: Ambient |
MDL Number:
MFCD00003423 CAS číslo: 10043-01-3 EINECS: 233-135-0 Merck Index: 14,366 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...