Diindium triselenide ≥99.99% (metals basis)
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Indium (III) selenide, Indiumselenide
88280.06
88280.14
88280.06EA
5530
CZK
88280.06
88280.14
Diindium triselenide ≥99.99% (metals basis)
Diindium triselenide
Purity: 99.99%
CAS No.: 12056-07-4
Molecular Formula: In2Se3
Formula Weight: 466.52
Physical Form: 3-6mm Pieces
Density: 5.67
Melting Point: ca 890°
MDL No.: MFCD00016147
Vzorec:
In₂Se₃ Bod tání: ∼ 890 °C Hustota: 5,67 Storage Temperature: Ambient |
MDL Number:
MFCD00016147 CAS číslo: 12056-07-4 EINECS: 235-016-9 UN: 3283 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...