Indium antimonide ≥99.99% (metals basis), Electronic Grade for the electronics industry
Dodavatel: Thermo Fisher Scientific
|
Varování
|
88460.09
88460.18
88460.04
88460.09EA
6320
CZK
88460.09
88460.18
88460.04
Indium antimonide ≥99.99% (metals basis), Electronic Grade for the electronics industry
Indium antimonide
Purity: 99.99%
CAS No.: 1312-41-0
Molecular Formula: InSb
Formula Weight: 236.57
Physical Form: Pieces
Density: 5.75 at m.p.
Melting Point: 535°
Merck Reference: 14,4948
UN No.: 1549
MDL No.: MFCD00016146
In semiconductor electronics, as an infrared detector and filter
Vzorec:
InSb Bod tání: 535 °C Hustota: 5,75 (m.p. Storage Temperature: Ambient |
MDL Number:
MFCD00016146 CAS číslo: 1312-41-0 EINECS: 215-192-3 UN: 1549 ADR: 6.1,III Merck Index: 14,4948 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...